Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPIL5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
| Antigen | PPIL5 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PPIL5 Polyclonal specifically detects PPIL5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PPIL5 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 122769 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LDDNELIQFPCKIGQLINLRFLSAARNKLPFLPSEFRNLSLEYLDLFGNTFEQPKVLPVIKLQAPLTLLESSART | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 4-1BBlrr, cyclophilin-like 5,4-1BB-mediated-signaling molecule, leucine rich repeat protein 1, LRR-1LRR-repeat protein 1, MGC20689,4-1BBLRR, peptidylprolyl isomerase (cyclophilin)-like 5,4-1BB-mediated signaling molecule, Peptidylprolyl isomerase-like 5, PPIL5 | |
| LRR1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title