Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPIL6 Antibody (OTI4F2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273578F
Description
PPIL6 Monoclonal specifically detects PPIL6 in Human samples. It is validated for Immunohistochemistry.Specifications
PPIL6 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PPIL6 | |
Full length recombinant protein of human PPIL6 (NP_775943) produced in HEK293T cells. | |
0.1 mL | |
285755.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4F2 | |
Immunohistochemistry | |
bA425D10.6, Cyclophilin-like protein PPIL6, dJ919F19.1, EC 5.2.1.8, MGC27054, MGC27056, MGC41939, peptidyl-prolyl cis-trans isomerase-like 6, peptidyl-prolyl cis-trans isomerase-like variant a, peptidylprolyl isomerase (cyclophilin)-like 6, PPIase, radial spoke 12 homolog, Rotamase PPIL6, RSPH12 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction