Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPIL6 Monoclonal Antibody (OTI4F2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504846S


Description
PPIL6 Monoclonal antibody specifically detects PPIL6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
PPIL6 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15773 | |
Mouse | |
Affinity chromatography | |
RUO | |
8079 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4F2 | |
Unconjugated | |
MLF2 | |
2900084F20Rik; bA425D10.6; Cyclophilin-like protein PPIL6; dJ919F19.1; peptidyl-prolyl cis-trans isomerase-like 6; peptidyl-prolyl cis-trans isomerase-like variant a; peptidylprolyl isomerase (cyclophilin)-like 6; peptidylprolyl isomerase like 6; PPIase; PPIL6; Probable inactive peptidyl-prolyl cis-trans isomerase-like 6; radial spoke 12 homolog; rotamase PPIL6; RSPH12 | |
Full length human recombinant protein of human MLF2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction