Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIP5K1/HISPPD2A Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501569033
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9033 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9033 Supplier Bethyl Laboratories, Inc Supplier No. A304921AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan

This gene encodes a dual functional inositol kinase. The encoded enzyme converts inositol hexakisphosphate to diphosphoinositol pentakisphosphate and diphosphoinositol pentakisphosphate to bis-diphosphoinositol tetrakisphosphate. This protein may be important for intracellular signaling pathways. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 15.

Specifications

Antigen PPIP5K1/HISPPD2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PPIP5K1
Gene Accession No. Q6PFW1
Gene Alias Diphosphoinositol pentakisphosphate kinase 1, HISPPD2A, histidine acid phosphatase domain containing 2A, histidine acid phosphatase domain-containing protein 2A, hsVIP1, inositol pyrophosphate synthase 1, insP6 and PP-IP5 kinase 1, IP6 kinase, IP6K, IPS1, KIAA0377, VIP1, VIP1 homolog
Gene Symbols PPIP5K1
Host Species Rabbit
Immunogen Between 1383 and 1433
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9677
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.