Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPIP5K2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570085
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
Inositol phosphates.
Specifications
PPIP5K2 | |
Polyclonal | |
Unconjugated | |
PPIP5K2 | |
AW555814; CFAP160; D330021B20; diphosphoinositol pentakisphosphate kinase 2; Hisppd1; histidine acid phosphatase domain containing 1; histidine acid phosphatase domain-containing protein 1; hsVIP2; inositol heptaphosphate kinase 2; inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; InsP6 and PP-IP5 kinase 2; IP7K2; KIAA0433; LOW QUALITY PROTEIN: inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; mKIAA0433; mmVIP2; PPIP5K2; VIP1 homolog 2; Vip2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
227399 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZQB6 | |
PPIP5K2 | |
synthetic peptide directed towards the middle region of human PPIP5K2 (aa 914-963). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction