Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPIP5K2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558390
Description
Immunogen sequence: LSVSSPEGTG TWLHYTSGVG TGRRRRRSGE QITSSPVSPK SLAFTSSIFG SWQQVVSENA NYLRTPRTLV EQKQNPTVGS HCA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 31%.
Inositol phosphates.
Specifications
PPIP5K2 | |
Polyclonal | |
Unconjugated | |
PPIP5K2 | |
AW555814; CFAP160; D330021B20; diphosphoinositol pentakisphosphate kinase 2; Hisppd1; histidine acid phosphatase domain containing 1; histidine acid phosphatase domain-containing protein 1; hsVIP2; inositol heptaphosphate kinase 2; inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; InsP6 and PP-IP5 kinase 2; IP7K2; KIAA0433; LOW QUALITY PROTEIN: inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; mKIAA0433; mmVIP2; PPIP5K2; VIP1 homolog 2; Vip2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23262 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43314 | |
PPIP5K2 | |
Recombinant protein corresponding to Human PPIP5K2. Recombinant protein control fragment (Product #RP-96885). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction