Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIP5K2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567031
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7031 Supplier Bethyl Laboratories, Inc Supplier No. A304166A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan

Inositol phosphates (IPs) and diphosphoinositol phosphates (PP-IPs), also known as inositol pyrophosphates, act as cell signaling molecules. HISPPD1 has both IP6 kinase (EC 2.7.4.21) and PP-IP5 (also called IP7) kinase (EC 2.7.4.24) activities that produce the high-energy pyrophosphates PP-IP5 and PP2-IP4 (also called IP8), respectively (Fridy et al., 2007 [PubMed 17690096]).

Specifications

Antigen PPIP5K2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PPIP5K2
Gene Accession No. O43314
Gene Alias HISPPD1, IP7K2, VIP2, VIP1 homolog 2, histidine acid phosphatase domain containing 1, histidine acid phosphatase domain-containing protein 1, hsVIP2, inositol heptaphosphate kinase 2, inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2, insP6 and PP-IP5 kinase
Gene Symbols PPIP5K2
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23262
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.