Learn More
PPIP5K2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304167A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Inositol phosphates (IPs) and diphosphoinositol phosphates (PP-IPs), also known as inositol pyrophosphates, act as cell signaling molecules. HISPPD1 has both IP6 kinase (EC 2.7.4.21) and PP-IP5 (also called IP7) kinase (EC 2.7.4.24) activities that produce the high-energy pyrophosphates PP-IP5 and PP2-IP4 (also called IP8), respectively (Fridy et al., 2007 [PubMed 17690096]).Specifications
PPIP5K2 | |
Polyclonal | |
Unconjugated | |
PPIP5K2 | |
HISPPD1, IP7K2, VIP2, VIP1 homolog 2, histidine acid phosphatase domain containing 1, histidine acid phosphatase domain-containing protein 1, hsVIP2, inositol heptaphosphate kinase 2, inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2, insP6 and PP-IP5 kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
227399, 23262 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O43314, Q6ZQB6 | |
PPIP5K2 | |
Between 925 and 975 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.