Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Periplakin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560020 Supplier Invitrogen™ Supplier No. PA560020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLAGLDKVAS DLDRQEKAIT GILRPPLEQG RAVQDSAERA KDLKNITNEL LRIEPEKTRS TAEGEAFIQA LPGSGTTPLL RTRVEDTNRK YEHLL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

The protein encoded by this gene is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Periplakin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ppl
Gene Accession No. O60437
Gene Alias 190 kDa paraneoplastic pemphigus antigen; 195 kDa cornified envelope precursor protein; 195kDa protein; AW553870; KIAA0568; MGC134872; Periplakin; Ppl
Gene Symbols Ppl
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPL. Recombinant protein control fragment (Product #RP-98660).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5493
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.