Learn More
Invitrogen™ PPM1A Monoclonal Antibody (7F12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517154
Description
MA5-17154 targets PPM1A in FACS, indirect ELISA, and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-17154 immunogen is purified recombinant fragment of human PPM1A (amino acids: 202-382) expressed in E. Coli. MA5-17154 detects PPM1A which has a predicted molecular weight of approximately 42.4kDa.
PPM1A (Protein phosphatase 1A) is also named as PPPM1A and belongs to the PP2C family. It is a serine/threonine protein phosphatase that is essential for regulating cellular stress responses in eukaryotes. PPM1A plays an important role in tumor suppression and that it negatively controls many signaling pathways which are critical for trophoblast invasion. This protein distributes in both the cytoplasm and nucleus of mammalian cells. It has 2 isoforms produced by alternative splicing.
Specifications
PPM1A | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P20650, P35813, P49443 | |
PPM1A | |
Purified recombinant fragment of human PPM1A (amino acids: 202-382) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
7F12 | |
Unconjugated | |
PPM1A | |
2310003C21Rik; 2900017D14Rik; AI427932; AU017636; IA; MMPa-2; Mpp alpha; MPPa-1; Pp2c1; PP2CA; PP2Calpha; PP2C-alpha; PPM1A; PPPM1A; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; protein phosphatase 1A, magnesium dependent, alpha isoform; protein phosphatase 2C isoform alpha; Protein phosphatase IA; Protein phosphatase type 1A (formely 2C) Mg-dependent alpha isoform; Protein phosphatase type 1A (formely 2C), Mg-dependent, alpha isoform; protein phosphatase, Mg2+/Mn2+ dependent 1A; protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
Mouse | |
Protein G | |
RUO | |
19042, 24666, 5494 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.