Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPM1B Monoclonal Antibody (OTI2H5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502811S


Description
PPM1B Monoclonal antibody specifically detects PPM1B in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
PPM1B | |
Monoclonal | |
0.72 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O94941 | |
Mouse | |
Affinity chromatography | |
RUO | |
22888 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Flow Cytometry, Western Blot | |
OTI2H5 | |
Unconjugated | |
Ubox5 | |
EGK_05287; Pp2c2; PP2CB; PP2CBETA; PP2C-beta; PP2C-beta-X; PPC2BETAX; PPM1B; Pppm1b; Protein phosphatase 1B; protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform; protein phosphatase 1B, magnesium dependent, beta isoform; protein phosphatase 2C isoform beta; protein phosphatase 2C-like protein; Protein phosphatase type 1B (formely 2C) Mg-dependent beta isoform; Protein phosphatase type 1B (formely 2C), Mg-dependent, beta isoform; protein phosphatase, Mg2+/Mn2+ dependent 1B; protein phosphatase, Mg2+/Mn2+ dependent, 1B; Ser/Thr protein phosphatase type 2C beta 2 isoform | |
Human recombinant protein fragment corresponding to amino acids 1-130 and 419-487 of human UBOX5 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction