Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPM1B Monoclonal Antibody (OTI3C2), TrueMAB™, OriGene
SDP

Catalog No. 502541294
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1294 30 μL
50-167-3825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1294 Supplier OriGene Supplier No. TA502812S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPM1B Monoclonal antibody specifically detects PPM1B in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PPM1B
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3C2
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PPM1B
Gene Accession No. O75688
Gene Alias EGK_05287; Pp2c2; PP2CB; PP2CBETA; PP2C-beta; PP2C-beta-X; PPC2BETAX; PPM1B; Pppm1b; Protein phosphatase 1B; protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform; protein phosphatase 1B, magnesium dependent, beta isoform; protein phosphatase 2C isoform beta; protein phosphatase 2C-like protein; Protein phosphatase type 1B (formely 2C) Mg-dependent beta isoform; Protein phosphatase type 1B (formely 2C), Mg-dependent, beta isoform; protein phosphatase, Mg2+/Mn2+ dependent 1B; protein phosphatase, Mg2+/Mn2+ dependent, 1B; Ser/Thr protein phosphatase type 2C beta 2 isoform
Host Species Mouse
Immunogen Full length human recombinant protein of human PPM1B produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5495
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.