Learn More
PPM1B Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300886A
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase has been shown to dephosphorylate cyclin-dependent kinases (CDKs), and thus may be involved in cell cycle control. Overexpression of this phosphatase is reported to cause cell-growth arrest or cell death. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional transcript variants have been described, but currently do not represent full-length sequences.Specifications
| PPM1B | |
| Polyclonal | |
| Unconjugated | |
| PPM1B | |
| PP2C-beta-X, PP2CB, PP2CBETA, PPC2BETAX, PP2C-beta, protein phosphatase 1B, protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform, protein phosphatase 2C isoform beta, protein phosphatase 2C-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5495 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O75688 | |
| PPM1B | |
| Between 400 and 450 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.