Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPM1D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572839
Description
PPM1D Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is a type IV membrane protein. It is present in the plasma membrane and intracellular vesicles. It may also be associated with the cytoskeleton. This protein may function in vesicle trafficking, membrane fusion, protein complex assembly and cell motility. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
PPM1D | |
Polyclonal | |
Unconjugated | |
PPM1D | |
AV338790; p53-induced protein phosphatase 1; PP2C delta; PP2CD; PP2C-delta; PPM1D; Protein phosphatase 1D; protein phosphatase 1D magnesium-dependent, delta isoform; protein phosphatase 2C delta isoform; protein phosphatase 2C isoform delta; Protein phosphatase magnesium-dependent 1 delta; protein phosphatase Mg2+/Mn2+ dependent 1D; protein phosphatase Wip1; protein phosphatase, Mg2+/Mn2+ dependent 1D; protein phosphatase, Mg2+/Mn2+ dependent, 1D; wild-type p53-induced phosphatase 1; WIP 1; WIP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
287585, 53892, 8493 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O15297, Q9QZ67 | |
PPM1D | |
PPM1D antibody for an 18 amino acid peptide near the carboxy terminus of human PPM1D. The immunogen is within the last 50 amino acids of PPM1D. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction