Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPM1E Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144162
Description
PPM1E Polyclonal Antibody for IHC (P), ELISA
Protein phosphatase that inactivates multifunctional CaM kinases such as CAMK4 and CAMK2. Dephosphorylates and inactivates PAK. May play a role in the inhibition of actin fiber stress breakdown and in morphological changes driven by TNK2/CDC42. Dephosphorylates PRKAA2.
Specifications
PPM1E | |
Polyclonal | |
Unconjugated | |
PPM1E | |
AW049266; B930008A12Rik; Ca(2+)/calmodulin-dependent protein kinase phosphatase N; calmodulin-dependent protein kinase phosphatase N; caMKN; CaMKP-N; caMKP-nucleus; Kiaa1072; mKIAA1072; nuclear calmodulin-dependent protein kinase phosphatase; partner of PIX 1; partner of PIXA; Partner of PIX-alpha; POPX1; PP2CE; PP2Ceta; PP2C-eta; PP2CH; PPM1E; PPM1M; Protein phosphatase 1E; protein phosphatase 1E (PP2C domain containing); protein phosphatase 1M; protein phosphatase 1M (PP2C domain containing); protein phosphatase 2C eta-2; protein phosphatase 2C isoform eta; protein phosphatase, Mg2+/Mn2+ dependent 1E; protein phosphatase, Mg2+/Mn2+ dependent 1M; protein phosphatase, Mg2+/Mn2+ dependent, 1E; protein phosphatase, Mg2+/Mn2+ dependent, 1M | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132160, 22843 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q8WY54, Q96MI6 | |
PPM1E, PPM1M | |
Recombinant Human Protein phosphatase 1M protein (1-247AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction