Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPM1E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553808 Supplier Invitrogen™ Supplier No. PA553808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVEGESLDLC LQQLYKYNCP SFLAAALARA TSDEVLQSDL SAHYIPKETD GTEGTVEIET VKLARSVFSK LHEI Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Protein phosphatase that inactivates multifunctional CaM kinases such as CAMK4 and CAMK2. Dephosphorylates and inactivates PAK. May play a role in the inhibition of actin fiber stress breakdown and in morphological changes driven by TNK2/CDC42. Dephosphorylates PRKAA2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPM1E
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPM1E
Gene Accession No. Q8WY54, Q96MI6
Gene Alias AW049266; B930008A12Rik; Ca(2+)/calmodulin-dependent protein kinase phosphatase N; calmodulin-dependent protein kinase phosphatase N; caMKN; CaMKP-N; caMKP-nucleus; Kiaa1072; mKIAA1072; nuclear calmodulin-dependent protein kinase phosphatase; partner of PIX 1; partner of PIXA; Partner of PIX-alpha; POPX1; PP2CE; PP2Ceta; PP2C-eta; PP2CH; PPM1E; PPM1M; Protein phosphatase 1E; protein phosphatase 1E (PP2C domain containing); protein phosphatase 1M; protein phosphatase 1M (PP2C domain containing); protein phosphatase 2C eta-2; protein phosphatase 2C isoform eta; protein phosphatase, Mg2+/Mn2+ dependent 1E; protein phosphatase, Mg2+/Mn2+ dependent 1M; protein phosphatase, Mg2+/Mn2+ dependent, 1E; protein phosphatase, Mg2+/Mn2+ dependent, 1M
Gene Symbols PPM1E, PPM1M
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPM1E. Recombinant protein control fragment (Product #RP-91581).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132160, 22843
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.