Learn More
PPM1G Mouse anti-Canine, Human, Clone: OTI1C12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502232

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.Specifications
PPM1G | |
Monoclonal | |
Unconjugated | |
PPM1G | |
PP2CG, PP2CGAMMA, PPP2CG | |
Mouse | |
Affinity Chromatography | |
RUO | |
475703, 5496 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15355 | |
PPM1G | |
Full length human recombit protein of human PPM1G produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.