Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPM1G Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | PPM1G |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-8695
|
Bethyl Laboratories, Inc
A300881AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.Specifications
| PPM1G | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Protein phosphatase 2C gamma isoform; PP2C-gamma; Protein phosphatase magnesium-dependent 1 gamma; Protein phosphatase 1C; PPM1C | |
| Between 500 and C-term | |
| Primary | |
| 4° C | |
| PPM1G |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O15355, Q61074 | |
| 14208, 5496 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title