Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPM1H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560656 Supplier Invitrogen™ Supplier No. PA560656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDQASCEVLT VKKKAGAVTS TPNRNSSKRR SSLPNGEGLQ LKENSESEGV SCHYWSLFDG H Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

Dephosphorylates CDKN1B at 'Thr-187', thus removing a signal for proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPM1H
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPM1H
Gene Accession No. Q9ULR3
Gene Alias A430075L18Rik; ARHCL1; C030002B11Rik; KIAA1157; NERPP-2C; neurite extension-related protein phosphatase related to PP2C; Ppm1h; Protein phosphatase 1H; protein phosphatase 1H (PP2C domain containing); protein phosphatase, Mg2+/Mn2+ dependent 1H; protein phosphatase, Mg2+/Mn2+ dependent, 1H; ras homolog gene family, member C like 1; URCC2
Gene Symbols PPM1H
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPM1H. Recombinant protein control fragment (Product #RP-102962).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57460
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.