Learn More
Invitrogen™ PPM1H Polyclonal Antibody

Description
PPM1H Polyclonal Antibody for Flow, IHC (P), Western Blot
Specifications
Specifications
| Antigen | PPM1H |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PPM1H |
| Gene Accession No. | Q3UYC0, Q9ULR3 |
| Gene Alias | A430075L18Rik; ARHCL1; C030002B11Rik; KIAA1157; NERPP-2C; neurite extension-related protein phosphatase related to PP2C; Ppm1h; Protein phosphatase 1H; protein phosphatase 1H (PP2C domain containing); protein phosphatase, Mg2+/Mn2+ dependent 1H; protein phosphatase, Mg2+/Mn2+ dependent, 1H; ras homolog gene family, member C like 1; URCC2 |
| Gene Symbols | PPM1H |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.