Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPM1J Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561137 Supplier Invitrogen™ Supplier No. PA561137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEVRVYDLTQ YEHCPDDVLV LGTDGLWDVT TDCEVAATVD RVLSAYEPND HSRYTALAQA LVLGARGTPR DRGWRLPNNK LGSGD Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene encodes the serine/threonine protein phosphatase. The mouse homolog of this gene apparently belongs to the protein phosphatase 2C family of genes. The exact function of this gene is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPM1J
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPM1J
Gene Accession No. Q5JR12
Gene Alias 2310008J22Rik; P PP2C-zeta; PP2CZ; PP2Czeta; PP2C-zeta; PPM1J; PPP2CZ; Protein phosphatase 1J; protein phosphatase 1J (PP2C domain containing); protein phosphatase 2a, catalytic subunit, zeta isoform; protein phosphatase 2C isoform zeta; protein phosphatase 2C zeta; protein phosphatase, Mg2+/Mn2+ dependent 1J; protein phosphatase, Mg2+/Mn2+ dependent, 1J
Gene Symbols PPM1J
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPM1J. Recombinant protein control fragment (Product #RP-94341).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.