Learn More
Invitrogen™ PPM1K Polyclonal Antibody

Description
Recommended positive controls: PPM1K tansfected 293T cell. Predicted reactivity: Mouse (92%), Rat (93%), Rhesus Monkey (98%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PPM1K |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PPM1K |
| Gene Accession No. | Q8N3J5 |
| Gene Alias | 2900063A19Rik; A930026L03Rik; BDP; branched-chain alpha-ketoacid dehydrogenase phosphatase; hPTMP; im:6912645; MSUDMV; PP2C domain-containing protein phosphatase 1K; PP2Ckappa; PP2C-kappa; PP2C-like mitochondrial protein; PP2C-like protein; PP2CM; PP2C-type mitochondrial phosphoprotein phosphatase; PPM1K; PPM1K protein; protein phosphatase 1K (PP2C domain containing); protein phosphatase 1K, mitochondrial; Protein phosphatase 2C isoform kappa; protein phosphatase 2C kappa; protein phosphatase, Mg2+/Mn2+ dependent 1K; protein phosphatase, Mg2+/Mn2+ dependent, 1K; PTMP; RGD1308501; UG0882E07; zgc:113207 |
| Gene Symbols | PPM1K |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.