Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPM1K Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554136 Supplier Invitrogen™ Supplier No. PA554136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PETKRIKLHH ADDSFLVLTT DGINFMVNSQ EICDFVNQCH DPNEAAHAVT EQAIQYGTED NSTAVVVPFG AWGKYKNSEI NFSFS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

PPM1K regulates the mitochondrial permeability transition pore and is essential for cellular survival and development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPM1K
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPM1K
Gene Accession No. Q8N3J5
Gene Alias 2900063A19Rik; A930026L03Rik; BDP; branched-chain alpha-ketoacid dehydrogenase phosphatase; hPTMP; im:6912645; MSUDMV; PP2C domain-containing protein phosphatase 1K; PP2Ckappa; PP2C-kappa; PP2C-like mitochondrial protein; PP2C-like protein; PP2CM; PP2C-type mitochondrial phosphoprotein phosphatase; PPM1K; PPM1K protein; protein phosphatase 1K (PP2C domain containing); protein phosphatase 1K, mitochondrial; Protein phosphatase 2C isoform kappa; protein phosphatase 2C kappa; protein phosphatase, Mg2+/Mn2+ dependent 1K; protein phosphatase, Mg2+/Mn2+ dependent, 1K; PTMP; RGD1308501; UG0882E07; zgc:113207
Gene Symbols PPM1K
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPM1K. Recombinant protein control fragment (Product #RP-91573).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.