Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPM1K Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554268
Description
Immunogen sequence: IGKRKENEDR FDFAQLTDEV LYFAVYDGHG GPAAADFCHT HMEKCIMDLL PKEKNLETLL TLAFLEIDKA FSSHAR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
PPM1K regulates the mitochondrial permeability transition pore and is essential for cellular survival and development.
Specifications
| PPM1K | |
| Polyclonal | |
| Unconjugated | |
| PPM1K | |
| 2900063A19Rik; A930026L03Rik; BDP; branched-chain alpha-ketoacid dehydrogenase phosphatase; hPTMP; im:6912645; MSUDMV; PP2C domain-containing protein phosphatase 1K; PP2Ckappa; PP2C-kappa; PP2C-like mitochondrial protein; PP2C-like protein; PP2CM; PP2C-type mitochondrial phosphoprotein phosphatase; PPM1K; PPM1K protein; protein phosphatase 1K (PP2C domain containing); protein phosphatase 1K, mitochondrial; Protein phosphatase 2C isoform kappa; protein phosphatase 2C kappa; protein phosphatase, Mg2+/Mn2+ dependent 1K; protein phosphatase, Mg2+/Mn2+ dependent, 1K; PTMP; RGD1308501; UG0882E07; zgc:113207 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 152926 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N3J5 | |
| PPM1K | |
| Recombinant protein corresponding to Human PPM1K. Recombinant protein control fragment (Product #RP-91574). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction