Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPM1K Polyclonal Antibody

Catalog No. PIPA595813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595813 Supplier Invitrogen™ Supplier No. PA595813

Rabbit Polyclonal Antibody

Positive Samples: A-431, Mouse heart, Mouse brain, Rat heart, Rat brain; Cellular Location: Mitochondrion matrix Immunogen sequence: KISLENVGCA SQIGKRKENE DRFDFAQLTD EVLYFAVYDG HGGPAAADFC HTHMEKCIMD LLPKEKNLET LLTLAFLEID KAFSSHARLS ADATLLTSGT TATVALLRDG IELVVASVGD SRAILCRKGK PMKLTIDHTP ERKDEKERIK KCGGFVAWNS LGQPHVNGRL AMTRSIGDLD LKTSGVIAEP ETKRIKLHHA DDSFLVLTTD GINFMVNSQE ICDFVNQCHD PNEAAHAVTE QAIQYGTEDN STAVVVPFGA WGK.

PPM1K regulates the mitochondrial permeability transition pore and is essential for cellular survival and development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPM1K
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PPM1K
Gene Accession No. Q8BXN7, Q8N3J5
Gene Alias 2900063A19Rik; A930026L03Rik; BDP; branched-chain alpha-ketoacid dehydrogenase phosphatase; hPTMP; im:6912645; MSUDMV; PP2C domain-containing protein phosphatase 1K; PP2Ckappa; PP2C-kappa; PP2C-like mitochondrial protein; PP2C-like protein; PP2CM; PP2C-type mitochondrial phosphoprotein phosphatase; PPM1K; PPM1K protein; protein phosphatase 1K (PP2C domain containing); protein phosphatase 1K, mitochondrial; Protein phosphatase 2C isoform kappa; protein phosphatase 2C kappa; protein phosphatase, Mg2+/Mn2+ dependent 1K; protein phosphatase, Mg2+/Mn2+ dependent, 1K; PTMP; RGD1308501; UG0882E07; zgc:113207
Gene Symbols PPM1K
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 89-351 of human PPM1K (NP_6897553).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152926, 243382, 312381
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.