Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPM1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554110
Description
Immunogen sequence: VLCDKDGNAI PLSHDHKPYQ LKERKRIKRA GGFISFNGSW RVQGILAMSR SLGDYPLKNL NVVIPDPDIL TF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PPM1L, or PP2CE, belongs to the PP2C group of serine/threonine phosphatases, which are distinguished from other phosphatases by their structure, absolute requirement for Mg(2+) or Mn(2+), and insensitivity to okadaic acid. PP2Cs regulate stress-activated protein kinase (SAPK.
Specifications
PPM1L | |
Polyclonal | |
Unconjugated | |
PPM1L | |
3222401G13; 5930404J21Rik; AI481720; AW045850; Kiaa4175; mKIAA4175; PP2C; PP2C epsilon; PP2CE; PP2Cepsilon; PP2C-epsilon; PPM1L; PPM1-LIKE; Ppp2ce; protein phosphatase 1 (formerly 2C)-like; Protein phosphatase 1L; protein phosphatase 1-like; protein phosphatase 2a, catalytic subunit, epsilon isoform; protein phosphatase 2C epsilon; Protein phosphatase 2C epsilon isoform; protein phosphatase 2C isoform epsilon; protein phosphatase, Mg2+/Mn2+ dependent 1L; protein phosphatase, Mg2+/Mn2+ dependent, 1L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
151742 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5SGD2 | |
PPM1L | |
Recombinant protein corresponding to Human PPM1L. Recombinant protein control fragment (Product #RP-91571). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction