Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPM1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554121 Supplier Invitrogen™ Supplier No. PA554121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLDKLQPEFM ILASDGLWDA FSNEEAVRFI KERLDEPHFG AKSIVLQSFY RGCPDNITVM VVKFRNSSK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

PPM1L, or PP2CE, belongs to the PP2C group of serine/threonine phosphatases, which are distinguished from other phosphatases by their structure, absolute requirement for Mg(2+) or Mn(2+), and insensitivity to okadaic acid. PP2Cs regulate stress-activated protein kinase (SAPK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPM1L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPM1L
Gene Accession No. Q5SGD2
Gene Alias 3222401G13; 5930404J21Rik; AI481720; AW045850; Kiaa4175; mKIAA4175; PP2C; PP2C epsilon; PP2CE; PP2Cepsilon; PP2C-epsilon; PPM1L; PPM1-LIKE; Ppp2ce; protein phosphatase 1 (formerly 2C)-like; Protein phosphatase 1L; protein phosphatase 1-like; protein phosphatase 2a, catalytic subunit, epsilon isoform; protein phosphatase 2C epsilon; Protein phosphatase 2C epsilon isoform; protein phosphatase 2C isoform epsilon; protein phosphatase, Mg2+/Mn2+ dependent 1L; protein phosphatase, Mg2+/Mn2+ dependent, 1L
Gene Symbols PPM1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPM1L. Recombinant protein control fragment (Product #RP-91572).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.