Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPME1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299601MFV610
Description
PPME1 Polyclonal antibody specifically detects PPME1 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
PPME1 | |
Polyclonal | |
Flow Cytometry, Flow (Intracellular) | |
EC 3.1.1, EC 3.1.1.-, FLJ22226, PME-1PME1, protein phosphatase methylesterase 1, protein phosphatase methylesterase-1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PPME1. | |
0.1 mL | |
Lipid and Metabolism | |
51400 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction