Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPME1 Antibody (OTI7F12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202240
Description
PPME1 Monoclonal specifically detects PPME1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PPME1 | |
Monoclonal | |
0.96 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
PPME1 | |
Full length human recombinant protein of human PPME1 (NP_057231) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI7F12 | |
Unconjugated | |
EC 3.1.1, EC 3.1.1.-, FLJ22226, PME-1PME1, protein phosphatase methylesterase 1, protein phosphatase methylesterase-1 | |
Mouse | |
42.3 kDa | |
0.1 mL | |
Lipid and Metabolism | |
51400 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction