Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPME1 Antibody (OTI7F12), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273583PCP
Description
PPME1 Monoclonal specifically detects PPME1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| PPME1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| PPME1 | |
| Full length recombinant protein of human PPME1 (NP_057231) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI7F12 | |
| Flow Cytometry | |
| EC 3.1.1, EC 3.1.1.-, FLJ22226, PME-1PME1, protein phosphatase methylesterase 1, protein phosphatase methylesterase-1 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 51400.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction