Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPME1 Mouse anti-Human, Clone: 29G3, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP25948750UL
Description
PPME1 Monoclonal specifically detects PPME1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
PPME1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
EC 3.1.1, EC 3.1.1.-, FLJ22226, PME-1PME1, protein phosphatase methylesterase 1, protein phosphatase methylesterase-1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
29G3 | |
Unconjugated | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
PPME1 | |
Recombinant human PPME1 (1-386aa) purified from E. coli | |
50 μL | |
Lipid and Metabolism | |
51400 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction