Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPNDS, Tocris Bioscience™
GREENER_CHOICE

Catalog No. p-4785372
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective P2X1 antagonist

Potent and selective P2X1 receptor antagonist (pKB = 7.43 in rat vas deferens). Up to 52-fold selective over P2Y1 receptors, and selective over ecto-nucleotidase, α1A-adrenoceptors, A1, A2B, H1 and M3 receptors.

Chemical Identifiers

CAS 1021868-77-8
Molecular Formula C18H11N4Na4O14PS2
Molecular Weight (g/mol) 694.353
InChI Key TXFLDKHOZFWVAC-UHFFFAOYSA-J
Synonym ppnds tetrasodium
PubChem CID 57369378
IUPAC Name tetrasodium;3-[2-[4-formyl-6-methyl-5-oxo-3-(phosphonatooxymethyl)pyridin-2-ylidene]hydrazinyl]-7-nitronaphthalene-1,5-disulfonate
SMILES CC1=NC(=NNC2=CC(=C3C=C(C=C(C3=C2)S(=O)(=O)[O-])[N+](=O)[O-])S(=O)(=O)[O-])C(=C(C1=O)C=O)COP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 50 mg
Formula Weight 694.36
Percent Purity >98%
Chemical Name or Material PPNDS
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.