Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPNDS, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective P2X1 receptor antagonist (pKB = 7.43 in rat vas deferens). Up to 52-fold selective over P2Y1 receptors, and selective over ecto-nucleotidase, α1A-adrenoceptors, A1, A2B, H1 and M3 receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 694.36 |
| Percent Purity | >98% |
| Chemical Name or Material | PPNDS |
Safety and Handling
| Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction