Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1CA Mouse anti-Human, Clone:1C11-C10-H5-E9, Abnova™

Catalog No. 89333881 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-881 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-881 Supplier Abnova Corporation Supplier No. MAB17659

Mouse monoclonal antibody raised against recombinant human PPP1CA.

The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Increased PP1 activity has been observed in the end stage of heart failure. Studies in both human and mice suggest that PP1 is an important regulator of cardiac function. Mouse studies also suggest that PP1 functions as a suppressor of learning and memory. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen PPP1CA
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1C11-C10-H5-E9
Conjugate Unconjugated
Description protein phosphatase 1, catalytic subunit, alpha isoform
Dilution Immunocytochemistry (1:300)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1000)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.1M Tris-Glycine, 150 mM NaCl, pH 7.4 (50% glycerol, 0.2% sodium azide)
Gene PPP1CA
Gene Alias MGC15877, MGC1674, PP-1A, PPP1A
Host Species Mouse
Immunogen Recombinant protein corresponding to a fragment of human PPP1CA from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5499
Test Specificity This antibody detects endogenous levels of PPP1A and does not cross-react with related proteins.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.