Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1CC Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572171
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2171 100 μL
50-157-2172 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2171 Supplier Bethyl Laboratories, Inc Supplier No. IHC00485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis, Bovine, Dog, Horse, Guinea pig_10141, Pig, Panda, Mo

Protein phosphatase-1 (PP1) is one of the major Ser/Thr protein phosphatases and is ubiquitous in eukaryotic cells. PP1 is localized to its site of action by interacting with targeting or regulatory proteins, a majority of which contains a primary docking site referred to as the RVXF/W motif. The catalytic subunit of PP1 exists as three isoforms, PP1a, PP1b, and PP1r. The PP1 catalytic subunits are small proteins of 37kDa, which are highly conserved, with their main differences falling in the C-term. The PP1 catalytic subunit exists in the cell in complex with a large number of targeting/regulatory subunits, thereby generating different enzyme forms that allow it to perform a diverse number of cellular functions.

Specifications

Antigen PPP1CC
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PPP1CC
Gene Accession No. P36873
Gene Alias MGC1675, MGC2870, PP2CG, PP2CGAMMA, PPP2CG
Gene Symbols PPP1CC
Host Species Rabbit
Immunogen Between 280 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5501
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.