Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563345 Supplier Invitrogen™ Supplier No. PA563345
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMDTASLEPG ALDAKPVESP GDPNQLTRKG RKRKSVTWPE EGKLREYFYF ELDETERVNV NKIKDFGEAA KREILSDRHA FETARRLSHD NMEEKVPWV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA40 and RPA16 are subunits of Pol I that associate with each other at an early stage of RNA polymerase I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III). RPA40, RPA16 and RPA135 encode the three subunits of RNA polymerase I, respectively. RPA194 is the largest subunit of RNA Pol I and is not a component of Pol II and Pol III.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R10
Gene Accession No. Q96QC0
Gene Alias 2610025H06Rik; Cat53; D17Ertd808e; Fb19; HLA-C adjacent transcript 53; HLA-C associated transcript 53; MHC class I region proline-rich protein CAT53; p99; Phosphatase 1 nuclear targeting subunit; Pnuts; PNUTSPP1R10; PP1-binding protein of 114 kDa; PP1R10; PPP1R10; Protein FB19; protein phosphatase 1 regulatory subunit 10; protein phosphatase 1, regulatory (inhibitor) subunit 10; protein phosphatase 1, regulatory subunit 10; protein phosphatase I; Protein PNUTS; putative protein phosphatase 1 nuclear targeting subunit; R111; Serine/threonine-protein phosphatase 1 regulatory subunit 10; testis tissue sperm-binding protein Li 67n
Gene Symbols PPP1R10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R10. Recombinant protein control fragment (Product #RP-103267).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5514
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.