Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566309
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566309 Supplier Invitrogen™ Supplier No. PA566309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSTESDEEEEE GCGHTHCVRG HRKGRRRATL Highest antigen sequence identity to the following orthologs - mouse 81%, rat 87%.

This gene encodes a specific inhibitor of protein phosphatase-1 (PP1) with a differential sensitivity toward the metal-independent and metal-dependent forms of PP1. The gene is located within the major histocompatibility complex class I region on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R11
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R11
Gene Accession No. O60927
Gene Alias 1500041B02Rik; AV117883; CFAP255; E3 ubiquitin-protein ligase PPP1R11; HCG V; HCGV; HCG-V; Hemochromatosis candidate gene V protein; hypothetical protein LOC550290; inhibitor-3; IPP3; Ppp1r11; protein phosphatase 1 regulatory inhibitor subunit 11; protein phosphatase 1 regulatory subunit 11; protein phosphatase 1, regulatory (inhibitor) subunit 11; protein phosphatase 1, regulatory (inhibitor) subunit 11, isoform 1; protein phosphatase inhibitor 3; T-complex testis expressed protein 5; t-complex testis expressed-5; t-complex testis-expressed 5; t-complex-associated-testis-expressed 5; TCTE5; Tctex5; Tctex-5; wu:fj64e04; zgc:110245
Gene Symbols PPP1R11
Host Species Rabbit
Immunogen Recombinant Human PPP1R11. Recombinant protein control fragment (Product #RP-106988).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.