Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555001 Supplier Invitrogen™ Supplier No. PA555001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTGINFWTKD EDETDGSEEV KETWHERLSR LESGGSNPTT SDSYGDRASA RARREAREAR LATLTSRVEE DSNRDYKKLY ESALTENQKL KTKLQEAQLE LADIKS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Myosin phosphatase is a protein complex comprised of three subunits: a catalytic subunit (PP1c-delta, protein phosphatase 1, catalytic subunit delta), a large regulatory subunit (MYPT, myosin phosphatase target) and small regulatory subunit (sm-M20). Two isoforms of MYPT have been isolated--MYPT1 and MYPT2, the first of which is widely expressed, and the second of which may be specific to heart, skeletal muscle, and brain. Each of the MYPT isoforms functions to bind PP1c-delta and increase phosphatase activity. This locus encodes both MYTP2 and M20. Alternatively spliced transcript variants encoding different isoforms have been identified. Related pseudogenes have been defined on the Y chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R12B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R12B
Gene Accession No. O60237
Gene Alias 1810037O03Rik; 9530009M10Rik; AI132431; Myosin phosphatase target subunit 2; myosin phosphatase, target subunit 2; myosin phosphatase-targeting subunit 2; MYPT2; PP1bp55; PPP1R12B; Protein phosphatase 1 regulatory subunit 12B; protein phosphatase 1, regulatory (inhibitor) subunit 12B; protein phosphatase 1, regulatory subunit 12B
Gene Symbols PPP1R12B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R12B. Recombinant protein control fragment (Product #RP-94025).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.