Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R12C Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144245
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144245 Supplier Invitrogen™ Supplier No. PA5144245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP1R12C Polyclonal Antibody for IHC (P), ICC/IF, ELISA

The gene encodes a subunit of myosin phosphatase. The encoded protein regulates the catalytic activity of protein phosphatase 1 delta and assembly of the actin cytoskeleton. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R12C
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene PPP1R12C
Gene Accession No. Q9BZL4
Gene Alias LENG3; leukocyte receptor cluster (LRC) encoded novel gene 3; leukocyte receptor cluster (LRC) member 3; MBS85; myosin-binding subunit 85; p84; p85; PPP1R12C; protein phosphatase 1 myosin-binding subunit of 85 kDa; Protein phosphatase 1 myosin-binding subunit p85; protein phosphatase 1 regulatory subunit 12C; protein phosphatase 1, regulatory (inhibitor) subunit 12C; protein phosphatase 1, regulatory subunit 12C
Gene Symbols PPP1R12C
Host Species Rabbit
Immunogen Recombinant Human Protein phosphatase 1 regulatory subunit 12C protein (317-450AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54776
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.