Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R12C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560408 Supplier Invitrogen™ Supplier No. PA560408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIARYLLSHG ANIAAVNSDG DLPLDLAESD AMEGLLKAEI ARRGVDVEAA KRAEEELLLH DTRCWLNGGA MPEARHPRTG ASALHV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The gene encodes a subunit of myosin phosphatase. The encoded protein regulates the catalytic activity of protein phosphatase 1 delta and assembly of the actin cytoskeleton. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R12C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R12C
Gene Accession No. Q9BZL4
Gene Alias LENG3; leukocyte receptor cluster (LRC) encoded novel gene 3; leukocyte receptor cluster (LRC) member 3; MBS85; myosin-binding subunit 85; p84; p85; PPP1R12C; protein phosphatase 1 myosin-binding subunit of 85 kDa; Protein phosphatase 1 myosin-binding subunit p85; protein phosphatase 1 regulatory subunit 12C; protein phosphatase 1, regulatory (inhibitor) subunit 12C; protein phosphatase 1, regulatory subunit 12C
Gene Symbols PPP1R12C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R12C. Recombinant protein control fragment (Product #RP-99011).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54776
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.