Learn More
Invitrogen™ PPP1R13B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552289
Description
Immunogen sequence: PNIQKLLYQR FNTLAGGMEG TPFYQPSPSQ DFMGTLADVD NGNTNANGNL EELPPAQPTA PLPAEPAPSS DANDNELPSP EPEELICPQT THQTAEPAED NNNNVATVPT TEQIPSPVAE APSPGEEQVP PAPLPPASHP PATSTNK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
This gene encodes a member of the ASPP family of p53 interacting proteins. The protein contains four ankyrin repeats and an SH3 domain involved in protein-protein interactions. ASPP proteins are required for the induction of apoptosis by p53-family proteins. They promote DNA binding and transactivation of p53-family proteins on the promoters of proapoptotic genes. Expression of this gene is regulated by the E2F transcription factor.
Specifications
PPP1R13B | |
Polyclonal | |
Unconjugated | |
PPP1R13B | |
AI449786; Apoptosis-stimulating of p53 protein 1; apoptosis-stimulating protein of p53, 1; ASPP1; AW545810; KIAA0771; p53BP2-like; p85; Ppp1r13b; Protein phosphatase 1 regulatory subunit 13B; protein phosphatase 1, regulatory (inhibitor) subunit 13B; protein phosphatase 1, regulatory subunit 13B; Tp53bp2; transformation related protein 53 binding protein 2; Trp53bp2; tumor protein p53 binding protein, 2; tumor protein p53-binding protein, 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21981, 23368, 314465 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q62415, Q96KQ4 | |
PPP1R13B | |
Recombinant protein corresponding to Human PPP1R13B. Recombinant protein control fragment (Product #RP-88994). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.