Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R13B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100703 Supplier Invitrogen™ Supplier No. PA5100703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ASPP1.

This gene encodes a member of the ASPP family of p53 interacting proteins. The protein contains four ankyrin repeats and an SH3 domain involved in protein-protein interactions. ASPP proteins are required for the induction of apoptosis by p53-family proteins. They promote DNA binding and transactivation of p53-family proteins on the promoters of proapoptotic genes. Expression of this gene is regulated by the E2F transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R13B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP1R13B
Gene Accession No. Q62415, Q96KQ4
Gene Alias AI449786; Apoptosis-stimulating of p53 protein 1; apoptosis-stimulating protein of p53, 1; ASPP1; AW545810; KIAA0771; p53BP2-like; p85; Ppp1r13b; Protein phosphatase 1 regulatory subunit 13B; protein phosphatase 1, regulatory (inhibitor) subunit 13B; protein phosphatase 1, regulatory subunit 13B; Tp53bp2; transformation related protein 53 binding protein 2; Trp53bp2; tumor protein p53 binding protein, 2; tumor protein p53-binding protein, 2
Gene Symbols PPP1R13B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP1R13B(Accession Q96KQ4), corresponding to amino acid residues N768-E818.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21981, 23368
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.