Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R13L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559491 Supplier Invitrogen™ Supplier No. PA559491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSEAFQSARD FLDMNFQSLA MKHMDLKQME LDTAAAKVDE LTKQLESLWS DSPAPPGPQA GPPSRPPRYS SSSIPEPFGS RGSPRKAATD GADTPFGRSE SAPTLHPYSP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53), whereas ASPP1. and ASPP2. are activators of p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R13L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R13L
Gene Accession No. Q8WUF5
Gene Alias AV260376; IASPP; inhibitor of apoptosis stimulating protein of p53; inhibitor of ASPP protein; LOW QUALITY PROTEIN: relA-associated inhibitor; NFkB interacting protein 1; NFkB-interacting protein 1; Nkip1; PPP1R13BL; PPP1R13B-like protein; PPP1R13L; protein iASPP; protein phosphatase 1 regulatory subunit 13 like; protein phosphatase 1, regulatory (inhibitor) subunit 13 like; protein phosphatase 1, regulatory subunit 13 like; RAI; RAI4; RelA-associated inhibitor; relA-associated inhibitor; LOW QUALITY PROTEIN: relA-associated inhibitor; retinoic acid induced 4; wa3
Gene Symbols PPP1R13L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R13L. Recombinant protein control fragment (Product #RP-103899).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10848
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.