Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R14B Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562415
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2415 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2415 Supplier Bethyl Laboratories, Inc Supplier No. A302365AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Pig

Inhibitor of PPP1CA. Has over 50-fold higher inhibitory activity when phosphorylated (By similarity).

Specifications

Antigen PPP1R14B
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PPP1R14B
Gene Accession No. Q96C90
Gene Alias PHI-1, PLCB3N, PNG, SOM172, phospholipase C beta-3 neighboring gene protein, phospholipase C-beta-3 neighbouring gene protein, protein phosphatase 1 regulatory subunit 14B
Gene Symbols PPP1R14B
Host Species Rabbit
Immunogen Between 97 and 147
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26472
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.