Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R14C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549854
Description
The antibody detects endogenous levels of total PP1R14C protein.
Inhibitor of the PP1 regulatory subunit PPP1CA.
Specifications
PPP1R14C | |
Polyclonal | |
Unconjugated | |
PPP1R14C | |
6330514J04Rik; CPI17-like; Kepi; kinase C-enhanced PP1 inhibitor; kinase-enhanced PP1 inhibitor; NY-BR-81; PKC-potentiated PP1 inhibitory protein; Ppp1r14c; protein phosphatase 1 regulatory inhibitor subunit 14C; protein phosphatase 1 regulatory subunit 14C; protein phosphatase 1, regulatory (inhibitor) subunit 14C; serologically defined breast cancer antigen NY-BR-81 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171010, 76142, 81706 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8R4R9, Q8R4S0, Q8TAE6 | |
PPP1R14C | |
Synthesized peptide derived from internal of human PP14C (aa 51-100). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction