Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R14C Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144821 Supplier Invitrogen™ Supplier No. PA5144821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IDIDDLLDAD SDEERASKLQ EALVDCY Antigen sequence identity: Mouse 93%; Rat 96%.

Inhibitor of the PP1 regulatory subunit PPP1CA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R14C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R14C
Gene Accession No. Q8TAE6
Gene Alias 6330514J04Rik; CPI17-like; Kepi; kinase C-enhanced PP1 inhibitor; kinase-enhanced PP1 inhibitor; NY-BR-81; PKC-potentiated PP1 inhibitory protein; Ppp1r14c; protein phosphatase 1 regulatory inhibitor subunit 14C; protein phosphatase 1 regulatory subunit 14C; protein phosphatase 1, regulatory (inhibitor) subunit 14C; serologically defined breast cancer antigen NY-BR-81
Gene Symbols PPP1R14C
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 111-137. Recombinant protein control fragment (Product #RP-109775).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81706
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.