Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R15A Monoclonal Antibody (OTI1E1), TrueMAB™, OriGene
SDP

Catalog No. 502540364
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0364 30 μL
50-167-5073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0364 Supplier OriGene Supplier No. TA504359S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPP1R15A Monoclonal antibody specifically detects PPP1R15A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PPP1R15A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E1
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Alox15
Gene Accession No. P16050
Gene Alias 9630030H21; Gadd34; growth arrest and DNA damage-inducible protein GADD34; growth arrest and DNA-damage-inducible 34; Myd116; myeloid differentiation primary response gene 116; myeloid differentiation primary response protein MyD116; Myeloid differentiation primary response protein MyD116 homolog; Peg3; PEG-3; PPP1R15A; PR15A; Progression elevated gene 3 protein; protein phosphatase 1 regulatory subunit 15A; protein phosphatase 1, regulatory (inhibitor) subunit 15A; protein phosphatase 1, regulatory subunit 15A; RGD1624209
Host Species Mouse
Immunogen Full length human recombinant protein of human ALOX15 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.