Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP1R15A Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI3D12, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA504310
Description
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation.Specifications
| PPP1R15A | |
| Monoclonal | |
| 0.25 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75807, P17564, Q6IN02 | |
| Ppp1r15a | |
| Full length human recombit protein of human PPP1R15A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D12 | |
| Unconjugated | |
| PPP1R15A | |
| GADD34 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 171071, 17872, 23645, 476419 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction