Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R15B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545140 Supplier Invitrogen™ Supplier No. PA545140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SFCSVDPYNP QNFTATIQTA ARIVPEEPSD SEKDLSGKSD LENSSQSGSL Sequence homology: Cow: 92%; Dog: 91%; Human: 100%.

Maintains low levels of EIF2S1 phosphorylation in unstressed cells by promoting its dephosphorylation by PP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R15B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PPP1R15B
Gene Accession No. Q5SWA1
Gene Alias 1810033K10Rik; AI606441; C530022L24Rik; constitutive repressor of eIF2alpha phosphorylation; CReP; MSSGM2; Ppp1r15b; Protein phosphatase 1 regulatory subunit 15B; protein phosphatase 1, regulatory (inhibitor) subunit 15b; protein phosphatase 1, regulatory subunit 15B
Gene Symbols PPP1R15B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PPP1R15B (aa 480-529).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84919
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.