Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R15B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595045
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.
Maintains low levels of EIF2S1 phosphorylation in unstressed cells by promoting its dephosphorylation by PP1.
Specifications
PPP1R15B | |
Polyclonal | |
Unconjugated | |
PPP1R15B | |
1810033K10Rik; AI606441; C530022L24Rik; constitutive repressor of eIF2alpha phosphorylation; CReP; MSSGM2; Ppp1r15b; Protein phosphatase 1 regulatory subunit 15B; protein phosphatase 1, regulatory (inhibitor) subunit 15b; protein phosphatase 1, regulatory subunit 15B | |
Rabbit | |
Affinity chromatography | |
RUO | |
84919 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
Q5SWA1 | |
PPP1R15B | |
A synthetic peptide corresponding to a sequence at the N-terminus of human PPP1R15B (15-31aa RAGFRFWPPFFPRRSQA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction